Orders LinkContactUsLinkComponent
VWR®, 遠心分離機用チューブ、感光性
VWR®, 遠心分離機用チューブ、感光性
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, 遠心分離機用チューブ、感光性
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, 遠心分離機用チューブ、感光性
VWR®, 遠心分離機用チューブ、感光性
6 Options Available Below
Product Details & Documents
PP, high grade gamma resistant, conical, amber, with white HDPE plug style cap with deep sealing area.
  • Tubes and caps are autoclavable and freezable
  • Gamma irradiated
  • Engraved graduations at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • Longer length screw caps with sealing ring prevent any leakage
  • Available in 15 or 50 ml sizes and offered in a variety of packaging formats
  • Certified non cytotoxic, nuclease-free and pyrogen-free
  • Sustainable design based on virgin resins
  • Blocks 99% of light with wavelength less than 550 nm
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Designed to provide protection for light-sensitive samples.
Specifications for Product Family
Specifications
  • Catalog No:
  • 525-0600
  • 525-0599
  • 525-0598
  • 525-0603
  • 525-0602
  • 525-0601
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
  • 容量
    Closure type
    RCF max.
    Version
    Packed
    Availability
    Price
  • 15 ml
    Screw cap
    12500×g
    non sterile
    Bulk, case of 500
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0600
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 15 ml
    Screw cap
    12500×g
    sterile
    50/bag,10 bags/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0599
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 15 ml
    Screw cap
    12500×g
    sterile
    25/rack, 20 racks/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0598
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 50 ml
    Screw cap
    12500×g
    non sterile
    Bulk, case of 500
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0603
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 50 ml
    Screw cap
    12500×g
    sterile
    50/bag,10 bags/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0602
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 50 ml
    Screw cap
    12500×g
    sterile
    25/rack, 20 racks/case
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0601
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact