Glycerine ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Merck Index:13,04497
Boiling Pt:290 °C (1013 hPa)
Density:1,26 g/cm³
Flash Pt:177 °C
Storage Temperature:Ambient
EINECS:200-289-5
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.5%
- For Laboratory,Research,or Manufacturing Use:Passes Test
- ACS - Assay (C₃H₅(OH)₃) (by GC, corrected for water):≥99.5 %
- ACS - Acrolein and Glucose:Passes Test
- ACS - Chlorinated Compounds (as Cl):≤0.003 %
- ACS - Color (APHA):≤10
- ACS - Fatty Acid Esters (as Butyric Acid):≤0.05 %
- ACS - Heavy Metals (as Pb):≤2 ppm
- ACS - Neutrality:Passes Test
- ACS - Residue after Ignition:≤0.005 %
- Solubility (5 in 50):Passes Test
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Sulfate (SO₄):≤0.001 %
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.5 %