Orders LinkContactUsLinkComponent
VWR®, 絶縁ガウン
VWR®, 絶縁ガウン
VWR®, 絶縁ガウン
Supplier:  VWR Collection
Certificates
VWR®, 絶縁ガウン
4 Options Available Below
Product Details & Documents
These disposable gowns, with back fastening, are made of non woven, breathable SMS fabric. Ideal for use in healthcare environments and hospitals.
  • Long sleeves, knitted cuffs
  • Round neck
  • Ties on neck and waist
  • Latex-free
Weight: 45 or 60 g/m²
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8315
  • 113-8316
  • 113-8321
  • 113-8322
  • Closure type:
  • Back fastening
  • Back fastening
  • Back fastening
  • Back fastening
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • Medical|Clinical
  • Medical|Clinical
  • Medical|Clinical
  • Medical|Clinical
  • Weight:
  • 45 g/m²
  • 45 g/m²
  • 60 g/m²
  • 60 g/m²
Specifications
Product Family Options
45 g/m²
Product Information
  • Colour
    W×L
    Availability
    Price
  • White
    1150×1360 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8315
    Closure type
    Back fastening
    Gender
    Unisex
    Garment application
    Medical|Clinical
    Weight
    45 g/m²
  • Blue
    1150×1360 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8316
    Closure type
    Back fastening
    Gender
    Unisex
    Garment application
    Medical|Clinical
    Weight
    45 g/m²
60 g/m²
Product Information
  • Colour
    W×L
    Availability
    Price
  • White
    1150×1360 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8321
    Closure type
    Back fastening
    Gender
    Unisex
    Garment application
    Medical|Clinical
    Weight
    60 g/m²
  • Blue
    1150×1360 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8322
    Closure type
    Back fastening
    Gender
    Unisex
    Garment application
    Medical|Clinical
    Weight
    60 g/m²