J.T.Baker®, Test Strips, Ion Specific
Catalog # 4418-01
Supplier: Avantor
CAS Number:
Specifications
- Pk:100
- 説明:Test strips, for sulfite ion determination
- Seq:01
Specifications
About this item
Baker TESTSTRIPS are made of plastic with test paper that is impregnated with a colorimetric reagent for use in semi-quantitative analysis. Strips generally measure 6,35×19,05 mm.
Strips generally measure ¼×3¼". Packaged in easy-to-use plastic boxes, each containing 100 strips.
Packaging: Packaged in easy-to-use plastic boxes, each containing 100 strips.