Orders LinkContactUsLinkComponent
pHインジケータースティック、BAKER-pHIX
 :   Avantor
pHインジケータースティック、BAKER-pHIX
 :  Avantor
pHインジケータースティック、BAKER-pHIX
pHインジケータースティック、BAKER-pHIX
BAKER-pHIX indicator sticks provide fast, easy, accurate measurement of pH ranges in laboratory.
Universal range sticks have resolution of 1,0 pH unit. Intermediate range sticks have resolutions of 0,3 to 0,5 pH units and narrow range sticks offer resolutions of 0,2 to 0,4 pH units.
 : Packaged in easy to use plastic boxes containing 100 sticks.
Product Family Options
Narrow range
  • 説明
    pH range
    pH gradation
  • pH indicator sticks, BAKER-pHIX
    1,7 - 3,8
    0,3
    1 * 100 items
    1 * 100 items
  • pH indicator sticks, BAKER-pHIX
    3,6 - 6,1
    0,3
    1 * 100 items
    1 * 100 items
  • pH indicator sticks, BAKER-pHIX
    5,2 - 6,8
    0,2/0,3
    1 * 200 items
    1 * 200 items
  • pH indicator sticks, BAKER-pHIX
    6,0 - 7,7
    0,3/0,4
    1 * 100 items
    1 * 100 items
  • pH indicator sticks, BAKER-pHIX
    7,5 - 9,5
    0,2/0,4