J.T.Baker® BAKERBOND Speedisk® Narc-2, SPE Columns
8175-04
: Avantor
- 粒子サイズ:10 µm
- 容量:3 ml
- Seq:01
- Bed weight:50 mg
- Phase:Narc-2
- Pk:1
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimise work and final solvent volumes and improve performance. The built-in column guard filter and higher analyte concentration may reduce or eliminate pre-filtration and concentration steps.
- Run times up to nine times faster than conventional SPE columns
- Unique laminar configuration
- Silica based
- Extraction columns for drugs of abuse
General applications: Hydrophobic/basic analytes (cocaine, benzoylecgonine).
These SPE columns can be used in mixed mode.