Ultimate grade FBS offers the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology. Premium grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a wide variety of applications.
- 100% US origin
- Triple 0,1 μm sterile filtration
- Preservation of native, growth-promoting factors and minimisation of growth-inhibiting factors
- Independently tested by third party laboratories
- Fully traceable
Ultimate grade FBS (endotoxin ≤5 EU/ml; haemogloblin ≤25 mg/dl) offers the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology. Proprietary collection and production techniques provide additional features that elevate product quality, enhance traceability and improve regulatory compliance. This product is used by cell culturists who work with primary cell lines, in production environments and applications where regulatory or traceability concerns are paramount.
Premium grade FBS (endotoxin ≤5 EU/ml; haemogloblin ≤25 mg/dl) offers quality features, such as complete testing by independent third party laboratories and raw material traceability that elevates it above comparable products. This product is used by cell culturists who require a high quality, high performance product for a wide variety of applications.
- Catalog No:
- 97068-086
- 89510-194
- 97068-085
- 97068-091
- Grade:
- Premium
- Premium
- Premium
- Premium
- Endotoxin:
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- ≤5 EU/ml
- Haemoglobin:
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- Storage temperature:
- Frozen
- Frozen
- Frozen
- Frozen