VWR®, Water Sampling Bottles, Sterile
New Product
PET, virgin, food grade with PP caps.
- Different coloured caps for easy indentification of test
- Tamper-evident strip to ensure sampling integrity
- Full traceability, batch number and individual bottle number on each label
- Fully recyclable
- Cap diameter: 32 mm
Lightweight, durable bottles. Caps designed to shield neck from contamination. Choice of dosed and undosed bottles. Rectangular bottles for better storage with limited space.
Blue cap: For natural, non chlorinated water without sodium thiosulphate.
Red cap: For chlorinated water dosed with 20 mg/L sodium thiosulphate.
White cap: For chlorinated water dosed with 120 mg/L sodium thiosulphate.
Certificate of Processing 1000B-157 Certificate of Processing 350A-66 Certificate of Processing 500B-73.pdf Certificate of Processing Bottle 1000 ml Square PET Sterile Certificate of Processing Ergo 500 ml Square 500A77
Additional Product Details
More
Certificate Of Processing_VWRI331-0269_1000A-90 Certificate Of Processing_VWRI331-0273_1000B-173 CoC_VWR_PET_WATER_SAMPLING_BOTTLES_WITH_SCREW_DOMED_CAP COC_VWRI331-0268_500A-70 coc_VWRI331-0270_10961531
More
VWR Bottles PR40268 VWR® for sampling and sample transportation
Product Literature
VWR PET Water Bottles with Screw Domed Cap