Orders LinkContactUsLinkComponent
VWR®, 乳鉢、乳棒付き、深型
Supplier:   VWR Collection
VWR®, 乳鉢、乳棒付き、深型
Supplier:  VWR Collection
Certificates
VWR®, 乳鉢、乳棒付き、深型
VWR®, 乳鉢、乳棒付き、深型
8 Options Available Below
Product Details & Documents
Agate.
  • High quality with flawless grinding surface.
Product Family Options
Product Information
  • 容量
    高さ
    深さ
    内径
    直径
    Availability
    Price
  • 25 ml
    36 mm
    20 mm
    50 mm
    60 mm
    1 * 1 items
  • 40 ml
    42 mm
    25 mm
    55 mm
    70 mm
    1 * 1 items
  • 65 ml
    53 mm
    30 mm
    65 mm
    80 mm
    1 * 1 items
  • 100 ml
    56 mm
    33 mm
    70 mm
    90 mm
    1 * 1 items
  • 125 ml
    61 mm
    33 mm
    80 mm
    100 mm
    1 * 1 items
  • 160 ml
    65 mm
    40 mm
    90 mm
    110 mm
    1 * 1 items
  • 210 ml
    68 mm
    42 mm
    100 mm
    120 mm
    1 * 1 items
  • 250 ml
    72 mm
    45 mm
    110 mm
    130 mm