Orders LinkContactUsLinkComponent
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
   Sustainable Options Available
Supplier:   VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
Supplier:  VWR Collection
   Sustainable Options Available
Certificates
VWR®, Centrifuge Tubes, Conical Bottom, 5 ml
6 Options Available Below
Product Details & Documents
These 5 ml microtubes are made of transparent PP with a snap flat cap, easy to be opened and closed.
  • Made from high grade gamma resistance PP
  • Autoclavable at 121 °C and freezable to –80 °C
  • Tubes can be centrifuged to 25000×g RCF
  • Provide easy and legible mark area
  • Sterile version is sterilised by irradiation SAL10⁻⁶ (ISO11137)
Micro centrifuge tubes are mainly used for sample storage, operation and centrifugation.
Certifications: Manufactured in a Class 100 000 cleanroom environment. Manufactured under ISO 13485:2016 and ISO 9001:2015 quality management systems.
Specifications for Product Family
Specifications
  • Catalog No:
  • 525-0666
  • 525-0661
  • 525-0662
  • 525-0663
  • 525-0664
  • 525-0665
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Product Family Options
Product Information
  • 容量
    Colour
    Closure type
    Version
    Packed
    Availability
    Price
  • 5 ml
    Natural
    With attached lid
    Sterile
    20/bag
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0666
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 5 ml
    Blue
    With attached lid
    Non sterile
    200/bag
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0661
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 5 ml
    Yellow
    With attached lid
    Non sterile
    200/bag
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0662
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 5 ml
    Green
    With attached lid
    Non sterile
    200/bag
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0663
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 5 ml
    Rose
    With attached lid
    Non sterile
    200/bag
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0664
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 5 ml
    Black
    With attached lid
    Non sterile
    200/bag
    Log in for availability
    Log in for availability
    1 * 200 items
    1 * 200 items
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    525-0665
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact