VWR®, フィルターチップ、エクストラロング
UltraFine™ point, extra long, 10 µl, +, +, 10 racks, 96 each
Supplier: VWR Collection
VWR®, フィルターチップ、エクストラロング
Catalog # 732-1613
Supplier: VWR Collection
Jump to:
Product Details & Documents
Aerosol filter tips with extended length.
- Sustainable design based on virgin resins
- Supplied sterile, autoclavable at 122 °C
- RNase-/DNase-, human DNA-, endotoxin-, protease-, ATP- and BPA-free
納品情報: Supplied in hinged racks, racks are refillable.
Specifications
- CE-IVD:+
- Packed:10 racks, 96 each
- Sterile:+
- 容量:10 µl
- Tip style:UltraFine™ point, extra long
- Cat. No.:732-1613
- Environmentally Preferable:Low Manufacturing Impact|Sustainable Packaging