Digital Solutions
Applications
Support
About
Japan
Orders Link
ContactUsLinkComponent
温度プローブ
VWR®, 超高精度プローブ(PT)
VWR®, 超高精度プローブ(PT)
:
VWR Collection
VWR®, 超高精度プローブ(PT)
:
VWR Collection
VWR®, 超高精度プローブ(PT)
VWR® for Temperature Measurement and Recording
Product Family Options
説明
Range
直径×長さ
620-1944
Immersion and air probe, Pt100, PTX 341, with plastic handle, PVC cable (1 m) and DIN connection
−50…+400 °C
3×150 mm
1 * 1 items
-
+
1 * 1 items
-
+
620-1949
Screw probe for deep-frozen products, PTZ 101 - SC, cable (1 m), Phono connection
−70…+200 °C
6×95 mm
-
+
-
+
VWR®, 遠心分離機用チューブ
#: 525-0640
: VWR Collection
VWR®, Transfer Pipettes, High Performance
#: 612-1684
: VWR Collection