Orders LinkContactUsLinkComponent
VWR®, pH測定用低流量セル
Low flow cell 200 µl, acrylic
  662-0457
VWR®, pH測定用低流量セル
  662-0457
VWR®, pH測定用低流量セル
VWR®, pH測定用低流量セル
Low flow cell 200 µl, acrylic
1 * 1 items
VWR® low flow cells are designed for use in a variety of instrumental techniques such as FIA, HPLC and IEF. They are also used to enable a continuous monitoring of the pH at low volume flows in bench scale chemical systems. They are supplied with inlet and outlet fittings sized for standard ¹/₁₆' or 1,6 mm OD tubing.
  • Designed for clean, sweeping flow at low volume
  • Designed for use with VWR® large surface flat tip electrode Cat. No. 662-0456
  • 説明:
    Low flow cell 200 µl, acrylic
  • Seq:
    32