Deep well storage plates, square well, Riplate® SW
613-1043
: RITTER
- Recommended working volume:50 µl – 2,4 ml
- Sterile:-
- 説明:96-well plates, PP, riplate® SW, 2 ml
- Pk:20
- Total volume:2,5 ml
Riplate® SW (= square wells), manufactured from pure polypropylene, have been developed for optimal use of capacity when it comes to sample storage.
- SBS format
- Thin-walled, parallel cavities ensure a maximum of contamination-free sample storage per well
- Alphanumeric marking for easy identification
- Suitable for sealing with cap mats, as well as adhesive film or heat sealing