Orders LinkContactUsLinkComponent
VWR®, ブレンダーバッグ
VWR®, ブレンダーバッグ
VWR®, ブレンダーバッグ
Supplier:  VWR Collection
Certificates
VWR®, ブレンダーバッグ
VWR®, ブレンダーバッグ
7 Options Available Below
Product Details & Documents
Wide choice of PE sterile bags from 80 to 3500 ml capacity. Fit all conventional laboratory blenders, adapted to all samples.
  • Bags with filters are mainly dedicated to fibrous samples
  • Bags with full filter for pasty samples
  • Plain bags for standard uses
  • No risk of cross-contamination
Absence of heavy material : raw material is compliant with the EC/94/62 directive.
Food contact: raw material is compliant with regulation EC N°1935/2004
Based on ISO 11137: Sterilisation of health care products SAL10-3
Certifications: Absence of heavy material: Raw material is compliant with the EC/94/62 directive
Food contact: Raw material is compliant with regulation EC No. 1935/2004
Based on ISO 11137: Sterilisation of healthcare products SAL 10⁻³
Product Family Options
Product Information
  • 説明
    容量
    Packed
    W×L
    Availability
    Price
  • Plain bags
    80 ml
    60 bags of 25
    105×150 mm
    Log in for availability
    Log in for availability
    1 * 1.500 items
    1 * 1.500 items
  • Plain bags
    400 ml
    20 bags of 25
    300×190 mm
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • Plain bags
    400 ml
    10 bags of 50
    300×190 mm
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • Plain bags
    3500 ml
    20 bags of 25
    380×510 mm
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • Bags with lateral filter
    400 ml
    10 bags of 50
    300×190 mm
    Log in for availability
    Log in for availability
    1 * 500 items
    1 * 500 items
  • Bags with lateral filter
    400 ml
    60 bags of 25
    300×190 mm
    Log in for availability
    Log in for availability
    1 * 1.500 items
    1 * 1.500 items
  • Bags with full page filter
    400 ml
    10 bags of 50
    300×190 mm