Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Holmium perchlorate solution
Holmium perchlorate solution Reag. Ph. Eur. 1043101
Holmium perchlorate solution Reag. Ph. Eur. 1043101
Holmium perchlorate solution Reag. Ph. Eur. 1043101
Catalog # 87841.290
Supplier:
VWR Chemicals
CAS Number:
Certificates
Expand image
Holmium perchlorate solution Reag. Ph. Eur. 1043101
Catalog # 87841.290
Supplier:
VWR Chemicals
CAS Number:
Certificates
Specifications
パックタイプ:
Plastic bottle
Pk:
1 L
Seq:
5037
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation