VWR®, ESDオーバーオール、フード付き
Supplier: VWR Collection
About this item
These breathable overalls are made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Elasticated hood
- Zip closure
- Elasticated cuffs, ankles and waist
- Anti-static properties
Weight: 50 g/m²
Colour: White
Packaging: 1 piece per bag, 50 bags per case.
6 Options Available Below
Product Details & Documents
These breathable overalls are made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
- Elasticated hood
- Zip closure
- Elasticated cuffs, ankles and waist
- Anti-static properties
Weight: 50 g/m²
Colour: White
Packaging: 1 piece per bag, 50 bags per case.
Documents
Food contact certificate F
Additional Product Details
TDS ESD Overalls, SPP, With Hood
Datasheet
VWR® Non Regulated Disposables for Production
Product Literature
Specifications for Product Family
Specifications
- Catalog No:
- 113-8305
- 113-8306
- 113-8307
- 113-8308
- 113-8309
- 113-8310
- Anti-static:
- +
- +
- +
- +
- +
- +
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Garment application:
- General Purpose|Industrial
- General Purpose|Industrial
- General Purpose|Industrial
- General Purpose|Industrial
- General Purpose|Industrial
- General Purpose|Industrial
- Weight:
- 50 g/m²
- 50 g/m²
- 50 g/m²
- 50 g/m²
- 50 g/m²
- 50 g/m²
Specifications
Product Family Options
Product Information
- Colour衣服のサイズAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-8305Anti-static+Cleanroom class100/ISO 5Garment applicationGeneral Purpose|IndustrialWeight50 g/m² - Specifications:
More
Specifications
Cat. No.113-8306Anti-static+Cleanroom class100/ISO 5Garment applicationGeneral Purpose|IndustrialWeight50 g/m² - Specifications:
More
Specifications
Cat. No.113-8307Anti-static+Cleanroom class100/ISO 5Garment applicationGeneral Purpose|IndustrialWeight50 g/m² - Specifications:
More
Specifications
Cat. No.113-8308Anti-static+Cleanroom class100/ISO 5Garment applicationGeneral Purpose|IndustrialWeight50 g/m² - Specifications:
More
Specifications
Cat. No.113-8309Anti-static+Cleanroom class100/ISO 5Garment applicationGeneral Purpose|IndustrialWeight50 g/m² - Specifications:
More
Specifications
Cat. No.113-8310Anti-static+Cleanroom class100/ISO 5Garment applicationGeneral Purpose|IndustrialWeight50 g/m²