Orders LinkContactUsLinkComponent
VWR®, 実験用白衣、SPP、使い捨て
VWR®, 実験用白衣、SPP、使い捨て
VWR®, 実験用白衣、SPP、使い捨て
Supplier:  VWR Collection
Certificates
VWR®, 実験用白衣、SPP、使い捨て
7 Options Available Below
Product Details & Documents
This disposable coat is made of non woven, laminated, SPP fabric.
  • Tailored collar
  • Press stud closure and open cuffs
  • Two pockets
Suitable for use in ISO Class 5 + 6 cleanroom environments.
Weight: 45/30 g/m²
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8297
  • 113-8298
  • 113-8299
  • 113-8302
  • 113-8303
  • 113-8304
  • 113-8759
  • Closure type:
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Buttons
  • Pockets:
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
Specifications
Product Family Options
Product Information
  • 説明
    Colour
    衣服のサイズ
    Availability
    Price
  • SPP 45 g/m²
    White
    M
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8297
    Closure type
    Buttons
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
  • SPP 45 g/m²
    White
    L
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8298
    Closure type
    Buttons
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
  • SPP 45 g/m²
    White
    XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8299
    Closure type
    Buttons
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
  • SPP 45 g/m²
    White
    XXL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8302
    Closure type
    Buttons
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
  • SPP 45 g/m²
    White
    3XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8303
    Closure type
    Buttons
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
  • SPP 45 g/m²
    White
    4XL
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8304
    Closure type
    Buttons
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
  • SPP 30 g/m²
    Blue
    XL
    Log in for availability
    Log in for availability
    1 * 30 items
    1 * 30 items
    Specifications:
    More
    Specifications
    Cat. No.
    113-8759
    Closure type
    Buttons
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory