VWR® Q Path®, 保管庫、引き出し2段
Catalog # 631-1072
Supplier: VWR Collection
Specifications
- 説明:Organiser with 2 metal drawers
- Seq:03
- Pk:1
Specifications
About this item
Storage organiser with two metal drawers to store embedding cassettes.
- For 2×700 cassettes
- Weight: 16 kg