Orders LinkContactUsLinkComponent
VWR®, 研削ジャー
Supplier:   VWR Collection
VWR®, 研削ジャー
Supplier:  VWR Collection
Certificates
VWR®, 研削ジャー
16 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • 容量
    Material
    Max. feed size
    Sample volume
    Availability
    Price
  • 1,5 ml
    Stainless steel
    1 mm
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 5 ml
    Stainless steel
    2 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 10 ml
    Stainless steel
    4 mm
    4 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 25 ml
    Stainless steel
    6 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 35 ml
    Stainless steel
    6 mm
    15 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 50 ml
    Stainless steel
    8 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 1,5 ml
    Steel, hardened
    1 mm
    0,5 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 5 ml
    Steel, hardened
    2 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 10 ml
    Steel, hardened
    4 mm
    4 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 25 ml
    Steel, hardened
    6 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 35 ml
    Steel, hardened
    6 mm
    15 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 50 ml
    Steel, hardened
    8 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 50 ml
    PTFE
    8 mm
    20 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 25 ml
    Tungsten carbide
    6 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 25 ml
    Stainless steel, forensic applications
    6 mm
    10 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
  • 35 ml
    Zirconium oxide
    10 mm
    15 ml
    Log in for availability
    Log in for availability
    1 * 2 items
    1 * 2 items
Complementary Products
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 412-0167
    Beater
    Log in for availability
    1 * 1 items - $0.00