VWR®, ベンチ保護紙
White, 25 sheets/pack, 371×303 mm
Catalog # 631-1059
Supplier: VWR Collection
Jump to:
Product Details & Documents
Highly absorbent paper sheets for bench and surface protection.
- Dust-proof packaging
- Very soft
- Prevents scratches on surfaces
Specifications
- Packed:25 sheets/pack
- Colour:White
- 長さ×幅:371×303 mm