Orders LinkContactUsLinkComponent
VWR® Q Path®, スライドオーガナイザー
VWR® Q Path®, スライドオーガナイザー
Supplier:   VWR Collection
VWR® Q Path®, スライドオーガナイザー
Supplier:  VWR Collection
Certificates
VWR® Q Path®, スライドオーガナイザー
VWR® Q Path®, スライドオーガナイザー
2 Options Available Below
Product Details & Documents
These organisers enable the archiving of a large number of microscope slides.
  • Robust and stackable
W×D×H: 480×480×140 mm
Delivery information: Supplied with locking rack.
Documents
Product Family Options
Product Information
  • 説明
    Colour
    W×D×H
    No. of slides
    Availability
    Price
  • Metal organiser with PVC slide drawers (14)
    Green
    480×480×140 mm
    5500
    1 * 1 items
  • Metal organiser with metal slide drawers (14)
    Green
    480×480×140 mm
    5500
    1 * 1 items
Accessories
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 631-1227
    Locking rack for conventional slides
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 631-1073
    Lid for organiser
    Log in for availability
    1 * 1 items
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 631-1071
    Base for organiser