Orders LinkContactUsLinkComponent
VWR®, 試験管、フチなし
VWR®, 試験管、フチなし
VWR®, 試験管、フチなし
VWR®, 試験管、フチなし
VWR®, 試験管、フチなし
Polystyrene (PS), transparent or polypropylene (PP), translucent.
Universal tubes with round bottom for a wide range of applications.
Product Family Options
  • 容量
    長さ
    外径
    Material
    Type
  • 3 ml
    55 mm
    11,5 mm
    PS
    Non graduated
    1 * 6.000 items
    1 * 6.000 items
  • 3 ml
    55 mm
    11,5 mm
    PP
    Non graduated
    1 * 6.000 items
    1 * 6.000 items
  • 3 ml
    70 mm
    11 mm
    PS
    Non graduated
    1 * 6.000 items
    1 * 6.000 items
  • 5 ml
    75 mm
    12 mm
    PP
    Sorwall type
    1 * 4.000 items
    1 * 4.000 items
  • 5 ml
    75 mm
    12 mm
    PS
    Non graduated Sorwall type
    1 * 4.000 items
    1 * 4.000 items
  • 5 ml
    75 mm
    12 mm
    PS
    Non graduated Sorwall type
    1 * 2.000 items
    1 * 2.000 items
  • 5 ml
    75 mm
    13 mm
    PS
    Non graduated
    1 * 4.000 items
    1 * 4.000 items
  • 5 ml
    75 mm
    13 mm
    PP
    Non graduated
    1 * 4.000 items
    1 * 4.000 items
  • 10 ml
    100 mm
    16 mm
    PS
    Non graduated
    1 * 2.000 items
    1 * 2.000 items
  • 10 ml
    100 mm
    16 mm
    PP
    Graduated
    1 * 2.000 items
    1 * 2.000 items
  • 10 ml
    100 mm
    16 mm
    PS
    Graduated
    1 * 2.000 items
    1 * 2.000 items
Stoppers
  •  
    Colour
    Ø top
  • Natural
    16 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • White
    16 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Blue
    16 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Green
    16 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Red
    16 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • White
    12 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Natural
    13 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Green
    13 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Yellow
    13 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Red
    13 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Blue
    12 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Red
    12 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Green
    12 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Yellow
    12 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Blue
    13 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • White
    13 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00
    $0.00
    1 * 1.000 items
       $0.00
  •  
    Colour
    Ø top
  • Natural
    12 mm
    1 * 1.000 items
    1 * 1.000 items - $0.00