プロテイン染色、ブルーバンディット、VWR®
Protein stain for 50 mini gels, 1 L
Blue BANDit is a safe and environmentally friendly alternative to traditional Coomassie® Blue staining.
- Reduces handling of hazardous chemicals: No methanol or acetic acid required
- High sensitivity, quick, simple procedure
- Pk:1 L
- 説明:Protein stain for 50 mini gels
- Cat. No.:K217-1L
- Environmentally Preferable:Safer Human and Environmental Health