Aluminium oxide basic, powder, BAKER ANALYZED® Brockmann Activity Grade I for chromatography, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1344-28-1
:
Formula:Al₂O₃
MDL Number:MFCD00003424
:101.96 g/mol
Merck Index:13,00355
Boiling Pt:2980 °C (1013 hPa)
Melting Pt:2045 °C
Density:3,94 g/cm³ (20 °C)
EINECS:215-691-6
REACH:01-2119529248-35
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:9.3 - 10.1
- pH of 5% Slurry at 20°C:9.3 - 10.1
- Adsorption, mg o-nitroaniline/g:≥1.0