VWR® Basic, Frocks with Velcro®
About this item
VWR® Basic SPP frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
- Snap collar, raglan sleeves, VELCRO® closure, knitted cuffs, no pockets
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility that is lot controlled and validated through independent laboratory testing.
Colour: White
5 Options Available Below
Product Details & Documents
VWR® Basic SPP frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
- Snap collar, raglan sleeves, VELCRO® closure, knitted cuffs, no pockets
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility that is lot controlled and validated through independent laboratory testing.
Colour: White
Specifications for Product Family
Specifications
- Catalog No:
- 113-1991
- 113-1988
- 113-1989
- 113-1990
- 113-1992
- Closure type:
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Hook-and-loop / VELCRO®
- Pockets:
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Without pockets
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Weight:
- 48 g/m²
- 48 g/m²
- 48 g/m²
- 48 g/m²
- 48 g/m²
Specifications
Product Family Options
Product Information
- Colour衣服のサイズAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.113-1991Closure typeHook-and-loop / VELCRO®PocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratoryWeight48 g/m² - Specifications:
More
Specifications
Cat. No.113-1988Closure typeHook-and-loop / VELCRO®PocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratoryWeight48 g/m² - Specifications:
More
Specifications
Cat. No.113-1989Closure typeHook-and-loop / VELCRO®PocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratoryWeight48 g/m² - Specifications:
More
Specifications
Cat. No.113-1990Closure typeHook-and-loop / VELCRO®PocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratoryWeight48 g/m² - Specifications:
More
Specifications
Cat. No.113-1992Closure typeHook-and-loop / VELCRO®PocketsWithout pocketsGenderUnisexGarment applicationCleanroom|LaboratoryWeight48 g/m²