VWR® Stepper Pro, 反復利用型ピペット
Individual pipette, mechanical
Catalog # 613-2169
Supplier: VWR Collection
Jump to:
Product Details & Documents
This pipette is designed for the dispensing of contaminated fluids and volatile liquids. The acceptance of dispenser tips with plunger helps to avoid contamination by ascending aerosols in the tip guaranteeing long-term, secure use.
- Flexible: Pipette allows 12 tip sizes, 10 volume adjustments and 120 program steps
- Volume adjustable from 1 to 5500 µl
- Ideal for long dispensing series due to its low weight (85 g)
- Ergonomic design allows one handed volume selection, loading and dispensing of liquids
- Volume adjustments and dosing volumes for different tips are clearly visible on the dispenser
Documents
Specifications
- 説明:Individual pipette, mechanical
- Cat. No.:613-2169
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Environmentally Preferable:Low Manufacturing Impact