Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Japan
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VWR Red PTFE/White Silicone Septa, 11mm Crimp-top Aluminum cap, 5.5mm Centre Hole, 1.0mm thickness 100pcs/pack,50packs/case 1 * 5.000 items
Catalog # VWRA548-2752
Supplier:
VWR Collection
New Product
Expand image
VWR Red PTFE/White Silicone Septa, 11mm Crimp-top Aluminum cap, 5.5mm Centre Hole, 1.0mm thickness 100pcs/pack,50packs/case 1 * 5.000 items
Catalog # VWRA548-2752
Supplier:
VWR Collection
New Product
Related Information
Documentation
CoC VWR Red PTFE - White Silicone Septa
Documentation
Frequently Bought Together
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection