Order Entry
Japan
ContactUsLinkComponent
Converter from serial RS232 to ethernet 10M/100M, w/power supply 5VDC, dim. 66x70x25mm, for Balances 1 * 1 items
  611-5818
Converter from serial RS232 to ethernet 10M/100M, w/power supply 5VDC, dim. 66x70x25mm, for Balances 1 * 1 items
  611-5818
Frequently Bought Together