MEDIUM RATCHET TUBING CLAMP 1000/PK 1 * 1.000 items
Catalog # MFLX06833-00BLK
Supplier: Avantor Fluid Handling
About this item
Won't rust, corrode, or distort.
- Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
- Housing material:Polyoxymethylene (POM) Acetal
- Description:Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 1000/pack
- Seq:02
- Environmentally Preferable:
- Pinch valve tube size:1/8" to 1/2" OD
- Warranty:3 Months
- Pk:1.000
Frequently Bought Together