$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
ACE EXCEL 2 C18, 50 X 2.1 MM 1 * 1 items
EXL1010502UAN
:
Avantor
ACE EXCEL 2 C18, 50 X 2.1 MM 1 * 1 items
EXL1010502UAN
:
Avantor
Frequently Bought Together
Avantor® ACE® C18, HPLC Columns, 3 µm
: Avantor