Reagent Bottle with cap, Narrow mouth, Polypropylene, 8ml 1 * 12 items
Catalog # VWRA215-4567
Supplier: VWR Collection
Reagent Bottle with cap, Narrow mouth, Polypropylene, 8ml 1 * 12 items
Catalog # VWRA215-4567
Supplier: VWR Collection
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection