Order Entry
Japan
ContactUsLinkComponent
VWR Polypropylene Film, Sterile. Ideal for short-term storage, incubation/ELISA. Temperature Range: -20°C to +120°C. Secure seal minimizes evaporation and reduces aerosols upon removal. 1 * 100 items
Catalog # 391-0678
Supplier:  VWR Collection
VWR Polypropylene Film, Sterile. Ideal for short-term storage, incubation/ELISA. Temperature Range: -20°C to +120°C. Secure seal minimizes evaporation and reduces aerosols upon removal. 1 * 100 items
Catalog # 391-0678
Supplier:  VWR Collection
Frequently Bought Together