Order Entry
Japan
ContactUsLinkComponent
Masterflex® Drum Pump Inlet Tube, CPVC, 47" L; 1/Each
Masterflex® Drum Pump Inlet Tube, CPVC, 47" L; 1/Each
Catalog # MFLX70604-68
undefined
Masterflex® Drum Pump Inlet Tube, CPVC, 47" L; 1/Each
Catalog # MFLX70604-68
About this item
Safely transfer corrosive chemicals, flammable liquids, and non-corrosive media.
  • Heavy-duty, thick-walled construction makes tube more durable
  • Tube length 47"
Replacement tubes do not include barrel adapter and mouting bracket; order separately.
  • 説明:
    Drum Pump Tube Set, CPVC, 47"L
  • Seq:
    06
  • Environmentally Preferable:
  • Model:
    70604-68
  • Wetted materials:
    CPVC
  • Tube length, inlet:
    47"
  • Warranty:
    3 years
  • Pk:
    1
Frequently Bought Together