Masterflex® Flowmeter Tripod Base Stand for 32460-Series Flowmeters
MFLX32462-50
Easily mount flowmeters on tripod for easy viewing.
- Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
- Description:Tripod base Flowmeter Stand For 32460
- Seq:06
- Environmentally Preferable:
- Model:32462-50
- Pk:1
Frequently Bought Together