$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
Cole-Parmer Centrifuge Neutral Tube,15 mL, with Seperate Cap,STRL,500/Cs 1 * 1 items
Catalog # MFLX15978-41
Supplier:
Avantor Fluid Handling
Expand image
Cole-Parmer Centrifuge Neutral Tube,15 mL, with Seperate Cap,STRL,500/Cs 1 * 1 items
Catalog # MFLX15978-41
Supplier:
Avantor Fluid Handling
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals