$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Japan
ContactUsLinkComponent
BOTTLE HDPE 125ML AMBER WIDE MOUTH W/CO MARK 1 * 12 items
215-4476
:
VWR Collection
BOTTLE HDPE 125ML AMBER WIDE MOUTH W/CO MARK 1 * 12 items
215-4476
:
VWR Collection
VWRA215-4476_VDF05H24.pdf
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals