Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Pipette Controller, Classic

VWR®, Pipette Controller, Classic

Blue
Catalog # 612-6657
Supplier:   VWR Collection
VWR®, Pipette Controller, Classic
Catalog # 612-6657
Supplier:  VWR Collection
VWR®, Pipette Controller, Classic
VWR®, Pipette Controller, Classic

Blue

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Pipette controller for glass and plastic pipettes from 1 to 100 ml.

  • Speed of suction and dispensing controlled through the finger triggers
  • Fully charged has approximately 8 h of continuous use available
  • Low battery light illuminates when 1 hour's power remains

Offers a unique combination of features to give precision fingertip control in a classic streamlined design, suitable for users with large or small hands. Gravity dispense mode is designed for use with 'To Deliver' (TD) pipettes.

Specifications
  • Colour code:
    Blue
  • Seq:
    01