Order Entry
Japan
ContactUsLinkComponent
VWR FILTER UPPER CUP 0.22UM PVDF 250ML 1 * 24 items
Catalog # VWRU76008-600
Supplier:  VWR Collection
VWR FILTER UPPER CUP 0.22UM PVDF 250ML 1 * 24 items
Catalog # VWRU76008-600
Supplier:  VWR Collection
Frequently Bought Together