Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR® Basic, Frocks

VWR® Basic, Frocks

Blue, L, 104 cm
Catalog # 113-8998
Supplier:   VWR Collection
VWR® Basic, Frocks
Catalog # 113-8998
Supplier:  VWR Collection
VWR® Basic, Frocks
VWR® Basic, Frocks

Blue, L, 104 cm

Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.

  • Press stud fastening
  • Elastic wrists
  • Raglan sleeves
  • Free from natural rubber latex

Weight: 40 g/m²

Packaging: 1 piece per bag, 50 bags per case.

Specifications
  • Clothing Size:
    L
  • Colour:
    Blue
  • Length:
    104 cm
  • Seq:
    03
  • Cat. No.:
    113-8998
  • Garment application:
    General Purpose|Laboratory
  • Gender:
    Unisex
  • Pockets:
    Without pockets
  • Closure type:
    Press studs / snaps
  • Weight:
    40 g/m²