VWR® Basic, Frocks
White, L, 104 cm
Catalog # 113-8995
Supplier: VWR Collection
Jump to:
Product Details & Documents
These disposable frocks are manufactured from a strong, lightweight and breathable, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime and certain dry particulates in non hazardous environments.
- Press stud fastening
- Elastic wrists
- Raglan sleeves
- Free from natural rubber latex
Weight: 40 g/m²
: 1 piece per bag, 50 bags per case.
Specifications
- Clothing Size:L
- Colour:White
- Length:104 cm
- Seq:01
- Cat. No.:113-8995
- Garment application:General Purpose|Laboratory
- Gender:Unisex
- Pockets:Without pockets
- Closure type:Press studs / snaps
- Weight:40 g/m²



