Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, COD calibration standards

VWR®, COD calibration standards

COD standard solution, 500 mg/L, Plastic bottle, 500 mL
Catalog # 85064.260
Supplier:   VWR Chemicals
VWR®, COD calibration standards
Catalog # 85064.260
Supplier:  VWR Chemicals
CAS Number:  
VWR®, COD calibration standards
VWR®, COD calibration standards

COD standard solution, 500 mg/L, Plastic bottle, 500 mL

CAS Number:  
Price
$431.46
Log in for availability
1 * 500 mL
Product Details & Documents

These standards are ideal to use as control standards for COD measurements to verify that correct analysis for COD has taken place.

Achieving an acceptable result for a control standard will improve confidence in sample readings for this important environmental parameter.

Specifications

Specifications

  • Concentration:
    500 mg/L
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    COD standard solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.