Sodium hydroxide 2 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Cubetainer®, 4 L
Supplier: Avantor
Sodium hydroxide 2 N in aqueous solution, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # 5633-03
Supplier: Avantor
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Standardisation at 25°C traceable to NIST Standard Reference Material.
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:109 °C (1013 hPa)
- Density:≤1.11 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Documents
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:1.990 - 2.010
- Protect from air to avoid absorption of carbon dioxide.:≤5 ppm
- Normality:1.990 - 2.010
- Chloride (Cl):≤ 5 ppm
- ACS - Heavy Metals (as Pb):≤ 1 ppm
- Trace Impurities - Iron (Fe):≤ 0.5 ppm
Specifications
- Pack type:Cubetainer®
- Pk:4 L



