Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Racks for VLSB Shaking Baths

VWR®, Racks for VLSB Shaking Baths

Stainless steel test tube rack for 12 and 18 l baths, 21, 19 mm
Catalog # 462-0505
Supplier:   VWR Collection
VWR®, Racks for VLSB Shaking Baths
Catalog # 462-0505
Supplier:  VWR Collection
VWR®, Racks for VLSB Shaking Baths
VWR®, Racks for VLSB Shaking Baths

Stainless steel test tube rack for 12 and 18 l baths, 21, 19 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • For tube Ø:
    19 mm
  • No. of Holes:
    21
  • Description:
    Stainless steel test tube rack for 12 and 18 l baths
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.