Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, DNA loading buffers

VWR®, DNA loading buffers

Loading Dye Blue 5X, 5× 1 ml, 1 KIT
Catalog # 733-2575
Supplier:   VWR Chemicals
VWR®, DNA loading buffers
Catalog # 733-2575
Supplier:  VWR Chemicals
CAS Number:  
VWR®, DNA loading buffers
VWR®, DNA loading buffers

Loading Dye Blue 5X, 5× 1 ml, 1 KIT

CAS Number:  
Price
$137.30
Log in for availability
1 * 1 KIT
Product Details & Documents

These DNA loading dyes are used to load DNA samples to agarose or SDS DNA gels for gel electrophoresis.

Suitable for TAE, TBE, SB and LB electrophoresis buffers.

 : EC Regulation No. 1907/2006 (REACH).

 : Each kit contains 5×1 ml.

 : Store at –20 °C for in vitro use only.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Loading Dye Blue 5X
  • Pk Info:
    5× 1 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.