Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, Tissue Processing/Embedding Cassettes, Premium

VWR®, Tissue Processing/Embedding Cassettes, Premium

Tissue cassettes, Premium, Green
Catalog # VWRU18000-006
Supplier:   VWR Collection
VWR®, Tissue Processing/Embedding Cassettes, Premium
Catalog # VWRU18000-006
Supplier:  VWR Collection
VWR®, Tissue Processing/Embedding Cassettes, Premium
VWR®, Tissue Processing/Embedding Cassettes, Premium

Tissue cassettes, Premium, Green

Price
$620.72
Log in for availability
1 * 1.000 items
Product Details & Documents

These cassettes made of acetyl polymer feature a fluidic flow design for the optimisation of tissue processing and embedding.

  • Resistant to histological solvents
  • Chamber is 10% longer allowing for an increased specimen range
  • 45° angled print surface compatible with all automatic printing devices
  • Attached cover has a large labelling area and uses a frangible hinge
  • Unique design with vented side walls to maximise fluid exchange

The vented sides and rear wall design work exceptionally well with fatty, dense, and other difficult to process tissue samples.

Specifications
  • Colour:
    Green
  • Description:
    Tissue cassettes, Premium
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.