VWR®, Alkali Burettes, Automatic Self-Zeroing, Class A
50 ml, 0,10 ml, ±0,05 ml
VWRU17579-894
Precision-grade borosilicate glass burette assembly and receiving bottle.
- Fluoropolymer resin stopcock eliminates contamination from lubricants
- Prefilled with activated alumina, ready to use
- Easy to read white enamel graduations
Both the U-shaped and straight drying tubes are prefilled with activated alumina.
: Supplied with fluoropolymer resin stopcock, alumina filled drying tubes, and receiving bottle.
- Capacity:50 ml
- Seq:02
- Division:0,10 ml
- Tolerance:±0,05 ml