VWR® Basic Lab Coats, SMS, Disposable
White, S, 93,35 cm
VWRU414004-366
SMS lab coats are manufactured from a latex-free, multi-layer spunbonded-melt-blown-spunbonded (SMS) fabric that offers high tensile strength, while remaining soft, comfortable and breathable.
- Tapered collar, inset sleeves and snap closure
- Knitted cuffs and three pockets
- Fluid and particulate barrier
- Light chemical splash resistance
Colour: White or blue
- Clothing Size:S
- Colour:White
- Length:93,35 cm
- Seq:02
- Cat. No.:VWRU414004-366
- Garment application:Cleanroom|Laboratory
- Gender:Unisex
- Pockets:With pockets
- Closure type:Press studs / snaps



