VWR® Basic Cleanroom Lab Coats, SPP, Disposable
White, S, 100 cm
Catalog # VWRU414004-346
Supplier: VWR Collection
Jump to:
Product Details & Documents
VWR® Basic SPP lab coats are manufactured from a strong, lightweight and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in non hazardous environments.
- Tapered collar, inset sleeves, snap closure
- Elasticated or knitted cuffs, no pockets or three pockets
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility and lot controlled and validated through independent laboratory testing.
: 25 or 30 coats per carton.
Specifications
- Clothing Size:S
- Colour:White
- Length:100 cm
- Seq:15
- Cat. No.:VWRU414004-346
- Garment application:Cleanroom|Laboratory|Industrial
- Gender:Unisex
- Pockets:With pockets
- Closure type:Press studs / snaps
- Weight:45 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



