VWR® Basic Cleanroom Lab Coats, SPP, Disposable
White, XXL, 100 cm
Catalog # VWRU414004-343
Supplier: VWR Collection
Jump to:
Product Details & Documents
VWR® Basic SPP lab coats are manufactured from a strong, lightweight and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in non hazardous environments.
- Tapered collar, inset sleeves, snap closure
- Elasticated or knitted cuffs, no pockets or three pockets
- Lightweight and breathable first line barrier protection
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility and lot controlled and validated through independent laboratory testing.
Packaging: 25 or 30 coats per carton.
Specifications
- Clothing Size:XXL
- Colour:White
- Length:100 cm
- Seq:25
- Cat. No.:VWRU414004-343
- Garment application:Cleanroom|Laboratory|Industrial
- Gender:Unisex
- Pockets:With pockets
- Closure type:Press studs / snaps
- Weight:45 g/m²



